siRNA Details
| Gene Symbol: C030037F17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK021128.1 | - | 3 | 249 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728262.1 | - | 3 | 249 | Chr. 8 - 14745320 - 14747427 [-] on Build GRCm38 | |

C030037F17Rik
Genomic Map