siRNA Details
| Gene Symbol: C030037D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038058.1 | 4 | 881 | |||
| GenBank mRNA | AK021127.1 | - | 4 | 689 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C030037D09Rik| Gene Symbol: C030037D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038058.1 | 4 | 881 | |||
| GenBank mRNA | AK021127.1 | - | 4 | 689 | ||