siRNA Details
| Gene Symbol: C030037D09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038058.1 | 3 | 584 | |||
| GenBank mRNA | AK021127.1 | - | 3 | 392 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY728261.1 | - | 3 | 385 | Chr. 11 - 88718842 - 88728363 [+] on Build GRCm38 | |

C030037D09Rik
Genomic Map