siRNA Details
| Gene Symbol: Shkbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048719.1 | - | 2 | 263 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB650638.1 | - | 2 | 263 | Chr. 7 - 27354350 - 27355848 [+] on Build GRCm38 | |

Shkbp1
Genomic Map