siRNA Details
| Gene Symbol: Ube2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020443.1 | - | 68 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ube2d3| Gene Symbol: Ube2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020443.1 | - | 68 | |||