siRNA Details
| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044137.1 | - | 1, 2 | 48 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp280c| Gene Symbol: Zfp280c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK044137.1 | - | 1, 2 | 48 | ||