siRNA Details
| Gene Symbol: 9530085P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020655.1 | - | 1 | 111 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722315.1 | - | 1 | 109 | Chr. 8 - 121856980 - 121857600 [-] on Build GRCm38 | |

9530085P06Rik
Genomic Map