siRNA Details
| Gene Symbol: B430105G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046573.1 | - | 1614 | |||
| AK049886.1 | - | 1116 | ||||
| BB324198.2 | - | 678 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B430105G09Rik| Gene Symbol: B430105G09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK046573.1 | - | 1614 | |||
| AK049886.1 | - | 1116 | ||||
| BB324198.2 | - | 678 | ||||