siRNA Details
| Gene Symbol: Rab3gap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005623.1 | - | 1 | 187 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705821.1 | - | 1 | 187 | Chr. 1 - 127938379 - 127938882 [-] on Build GRCm38 | |

Rab3gap1
Genomic Map