siRNA Details
| Gene Symbol: 4930518J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005624.1 | - | 1 | 764 | ||
| AK015829.1 | - | 1 | 756 | |||
| AV209277.1 | - | 1 | 229 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930518J20Rik| Gene Symbol: 4930518J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005624.1 | - | 1 | 764 | ||
| AK015829.1 | - | 1 | 756 | |||
| AV209277.1 | - | 1 | 229 | |||