siRNA Details
| Gene Symbol: 4930518J20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK005624.1 | - | 1 | 453 | ||
| AK015829.1 | - | 1 | 445 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI595103.1 | - | 1 | 248 | Chr. 1 - 156303697 - 156304200 [+] on Build GRCm38 | |

4930518J20Rik
Genomic Map