siRNA Details
| Gene Symbol: 1110013I04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK003663.1 | - | 1 | 80 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY703926.1 | - | 1 | 81 | Chr. 8 - 112961448 - 112962560 [+] on Build GRCm38 | |

1110013I04Rik
Genomic Map