siRNA Details
| Gene Symbol: 4933416M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045846.2 | 2 | 274 | |||
| GenBank mRNA | AK016834.1 | - | 2 | 274 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716508.1 | - | 2 | 270 | Chr. 13 - 101254323 - 101293055 [+] on Build GRCm38 | |

4933416M06Rik
Genomic Map