siRNA Details
| Gene Symbol: 4933416M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045846.2 | 6 | 948 | |||
| GenBank mRNA | AK016834.1 | - | 6 | 948 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933416M06Rik| Gene Symbol: 4933416M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045846.2 | 6 | 948 | |||
| GenBank mRNA | AK016834.1 | - | 6 | 948 | ||