siRNA Details
| Gene Symbol: Gm16897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033564.1 | 5 | 2157 | |||
| GenBank mRNA | AK030377.1 | - | 5 | 2159 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16897| Gene Symbol: Gm16897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033564.1 | 5 | 2157 | |||
| GenBank mRNA | AK030377.1 | - | 5 | 2159 | ||