siRNA Details
| Gene Symbol: 2310001H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030376.1 | - | 1399 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310001H17Rik| Gene Symbol: 2310001H17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK030376.1 | - | 1399 | |||