siRNA Details
Gene Symbol: Dep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034614.1 | - | 1 | 1970 | ||
BB088198.2 | - | 1 | 448 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034614.1 | - | 1 | 1970 | ||
BB088198.2 | - | 1 | 448 |