siRNA Details
| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008044.1 | - | 1 | 660 | ||
| AK147721.1 | - | 1422 | ||||
| AK147792.1 | - | 1424 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mpc2| Gene Symbol: Mpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK008044.1 | - | 1 | 660 | ||
| AK147721.1 | - | 1422 | ||||
| AK147792.1 | - | 1424 | ||||