siRNA Details
| Gene Symbol: 2610005G05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011318.1 | - | 1 | 506 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV111739.1 | - | 1 | 117 | Chr. 16 - 26992291 - 26992571 [+] on Build GRCm38 | |

2610005G05Rik
Genomic Map