siRNA Details
| Gene Symbol: 4930583P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040612.1 | 3 | 459 | |||
| GenBank mRNA | AK016345.1 | - | 3 | 431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930583P06Rik| Gene Symbol: 4930583P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040612.1 | 3 | 459 | |||
| GenBank mRNA | AK016345.1 | - | 3 | 431 | ||