siRNA Details
| Gene Symbol: 4930583P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040612.1 | 1 | 125 | |||
| GenBank mRNA | AK016345.1 | - | 1 | 97 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA465385.1 | - | 1 | 128 | Chr. 2 - 124217689 - 124222360 [-] on Build GRCm38 | |

4930583P06Rik
Genomic Map