siRNA Details
| Gene Symbol: 4930583P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040612.1 | 1 | 302 | |||
| GenBank mRNA | AK016345.1 | - | 1 | 274 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930583P06Rik| Gene Symbol: 4930583P06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040612.1 | 1 | 302 | |||
| GenBank mRNA | AK016345.1 | - | 1 | 274 | ||