siRNA Details
| Gene Symbol: Gm21464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032382.1 | - | 3 | 1250 | ||
| AK086749.1 | - | 5 | 1422 | |||
| BC086315.1 | - | 4 | 1258 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm21464| Gene Symbol: Gm21464 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK032382.1 | - | 3 | 1250 | ||
| AK086749.1 | - | 5 | 1422 | |||
| BC086315.1 | - | 4 | 1258 | |||