siRNA Details
| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019679.1 | - | 2 | 1316 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

P2rx3| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019679.1 | - | 2 | 1316 | ||