siRNA Details
| Gene Symbol: 4930512P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK019678.1 | - | 1 | 223 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715322.1 | - | 1 | 223 | Chr. 3 - 116663732 - 116664155 [+] on Build GRCm38 | |

4930512P04Rik
Genomic Map