siRNA Details
| Gene Symbol: 4930511E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040761.1 | 5 | 841 | |||
| GenBank mRNA | AK019674.1 | - | 5 | 843 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930511E03Rik| Gene Symbol: 4930511E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040761.1 | 5 | 841 | |||
| GenBank mRNA | AK019674.1 | - | 5 | 843 | ||