siRNA Details
| Gene Symbol: 4930511E03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040761.1 | 4 | 464 | |||
| GenBank mRNA | AK019674.1 | - | 4 | 466 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715290.1 | - | 4 | 465 | Chr. 6 - 94943834 - 94946931 [+] on Build GRCm38 | |

4930511E03Rik
Genomic Map