siRNA Details
| Gene Symbol: 4930412L05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015122.1 | - | 5 | 560 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714668.1 | - | 5 | 560 | Chr. 4 - 53207279 - 53217561 [-] on Build GRCm38 | |

4930412L05Rik
Genomic Map