siRNA Details
| Gene Symbol: 4930413F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045883.2 | 3 | 371 | |||
| GenBank mRNA | AK015129.1 | - | 3 | 372 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY714675.1 | - | 3 | 372 | Chr. 15 - 34675073 - 34678728 [+] on Build GRCm38 | |

4930413F20Rik
Genomic Map