siRNA Details
| Gene Symbol: 4930413F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045883.2 | 4 | 855 | |||
| GenBank mRNA | AK015129.1 | - | 4 | 856 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930413F20Rik| Gene Symbol: 4930413F20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045883.2 | 4 | 855 | |||
| GenBank mRNA | AK015129.1 | - | 4 | 856 | ||