siRNA Details
| Gene Symbol: 4930413E20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK015128.1 | - | 1 | 490 | ||
| AK029610.1 | - | 1 | 490 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB614994.1 | - | 1 | 490 | Chr. 8 - 23535236 - 23535745 [+] on Build GRCm38 | |

4930413E20Rik
Genomic Map