siRNA Details
| Gene Symbol: 4933406J10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046004.1 | 4 | 563 | |||
| GenBank mRNA | AK016697.1 | - | 4 | 564 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BU945775.1 | - | 3 | 300 | Chr. 7 - 82743357 - 82754626 [-] on Build GRCm38 | |

4933406J10Rik
Genomic Map