siRNA Details
| Gene Symbol: B130024G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045850.1 | 9 | 1028 | |||
| GenBank mRNA | AK045070.1 | - | 9 | 1029 | ||
| BC070425.1 | - | 9 | 1260 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | NM_001004183.1 | - | 1029 | Chr. - - [-] on Build | ||

B130024G19Rik
Genomic Map