siRNA Details
| Gene Symbol: B130024G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045850.1 | 4 | 444 | |||
| GenBank mRNA | AK045070.1 | - | 4 | 445 | ||
| BC070425.1 | - | 4 | 671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

B130024G19Rik| Gene Symbol: B130024G19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045850.1 | 4 | 444 | |||
| GenBank mRNA | AK045070.1 | - | 4 | 445 | ||
| BC070425.1 | - | 4 | 671 | |||