siRNA Details
| Gene Symbol: 4930557F08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016160.1 | - | 1, 2 | 102 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715764.1 | - | 1, 2 | 98 | Chr. 15 - 15779996 - 15823426 [+] on Build GRCm38 | |

4930557F08Rik
Genomic Map