siRNA Details
| Gene Symbol: Gm15886 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016162.1 | - | 4, 5 | 465 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY715765.1 | - | 4, 5 | 465 | Chr. 3 - 103771973 - 103778109 [+] on Build GRCm38 | |

Gm15886
Genomic Map