siRNA Details
| Gene Symbol: 4833422M21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK016691.1 | - | 3 | 594 | ||
| AK084200.1 | - | 3 | 623 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY716377.1 | - | 3 | 594 | Chr. 15 - 96232368 - 96241789 [-] on Build GRCm38 | |

4833422M21Rik
Genomic Map