siRNA Details
| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043531.1 | - | 1649 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

H2afy| Gene Symbol: H2afy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043531.1 | - | 1649 | |||