siRNA Details
| Gene Symbol: D730050B12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046196.1 | 3 | 286 | |||
| GenBank mRNA | AK021357.1 | - | 3 | 287 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY730955.1 | - | 3 | 287 | Chr. 13 - 72809608 - 72816763 [-] on Build GRCm38 | |

D730050B12Rik
Genomic Map