siRNA Details
| Gene Symbol: Hotair | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047528.1 | 2 | 1622 | |||
| GenBank mRNA | AK035706.1 | - | 2 | 1406 | ||
| BB123558.2 | - | 1 | 91 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hotair| Gene Symbol: Hotair | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_047528.1 | 2 | 1622 | |||
| GenBank mRNA | AK035706.1 | - | 2 | 1406 | ||
| BB123558.2 | - | 1 | 91 | |||