siRNA Details
| Gene Symbol: 1500032F14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK018780.1 | - | 1 | 616 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AV031986.1 | - | 1 | 162 | Chr. 19 - 4256960 - 4257227 [-] on Build GRCm38 | |

1500032F14Rik
Genomic Map