siRNA Details
Gene Symbol: 5330431K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019926.1 | - | 1 | 1688 | ||
BG082526.2 | - | 1 | 451 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 5330431K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019926.1 | - | 1 | 1688 | ||
BG082526.2 | - | 1 | 451 |