siRNA Details
| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054399.1 | - | 1813 | |||
| AK149418.1 | - | 1004 | ||||
| BC048953.1 | - | 1789 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb20| Gene Symbol: Zbtb20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK054399.1 | - | 1813 | |||
| AK149418.1 | - | 1004 | ||||
| BC048953.1 | - | 1789 | ||||