siRNA Details
| Gene Symbol: Gm13944 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040368.1 | 3 | 581 | |||
| GenBank mRNA | AK078367.1 | - | 3 | 532 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY705307.1 | - | 3 | 551 | Chr. 2 - 77462677 - 77480801 [+] on Build GRCm38 | |

Gm13944
Genomic Map