siRNA Details
| Gene Symbol: Gm16740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108026.1 | 1 | 553 | |||
| GenBank mRNA | AK020406.1 | - | 1 | 554 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm16740| Gene Symbol: Gm16740 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108026.1 | 1 | 553 | |||
| GenBank mRNA | AK020406.1 | - | 1 | 554 | ||