siRNA Details
| Gene Symbol: 9430006E15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020405.1 | - | 1 | 253 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY721574.1 | - | 1 | 251 | Chr. 9 - 20687820 - 20688466 [+] on Build GRCm38 | |

9430006E15Rik
Genomic Map