siRNA Details
| Gene Symbol: 2010107E04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020400.1 | - | 1 | 122 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CA751014.1 | - | 1 | 494 | Chr. 12 - 111963401 - 111964176 [-] on Build GRCm38 | |

2010107E04Rik
Genomic Map