siRNA Details
Gene Symbol: 1700081H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040693.1 | 1 | 201 | |||
GenBank mRNA | AK006966.1 | - | 1 | 236 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700081H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040693.1 | 1 | 201 | |||
GenBank mRNA | AK006966.1 | - | 1 | 236 |