siRNA Details
| Gene Symbol: 1700021L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006219.1 | - | 1 | 588 | ||
| BC048567.1 | - | 1 | 563 | |||
| BC099561.1 | - | 1 | 398 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY706305.1 | - | 1 | 587 | Chr. 6 - 38433108 - 38433783 [-] on Build GRCm38 | |

1700021L22Rik
Genomic Map