siRNA Details
| Gene Symbol: Gm12292 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK036759.1 | - | 2 | 435 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB629752.1 | - | 2 | 435 | Chr. 11 - 64842379 - 64843753 [-] on Build GRCm38 | |

Gm12292
Genomic Map